Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butyl hexahydropyrrolo[3,4-b]pyrrole-5(1H)-carboxylate

Catalog No. CDS019398 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS019398 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS019398 Supplier Sigma Aldrich Supplier No. CDS019398100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Building block used as a substrate in phosphine-catalyzed domino sequences
  • This versatile compound can be used to synthesize a wide scope of spirocyclic benzofuranone scaffolds.asymmetric construction of spirocyclopentenebenzofuranone core structures via highly selective phosphine-catalyzed [3 + 2] cycloaddition reactions

Specifications

Molecular Formula C11H20N2O2
Linear Formula C11H20N2O2
MDL Number MFCD08235016
Quantity 100 mg
Molecular Weight (g/mol) 212.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.