Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl hydrogen tetradecanedioate | 234082-00-9 | 314.46 g/mol | C18H34O4
SDP

Catalog No. 5000210809
Encompass_Preferred

Tert-butyl hydrogen tetradecanedioate is a tert-butyl ester derivative used as a biochemical assay reagent and synthetic building block in research applications. It has CAS number 234082-00-9, molecular formula C18H34O4, and molecular weight 314.46 g/mol. Storage recommendations include room temperature storage for up to 3 years; in solvent, store at -80°C for 2 years or -20°C for 1 year.

  • CAS number: 234082-00-9
  • Molecular formula: C18H34O4
  • Molecular weight: 314.46 g/mol
  • Intended use: biochemical assay reagents
  • Storage: room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-002-10809 Supplier Medchemexpress LLC Supplier No. HYW07495350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tert-butyl hydrogen tetradecanedioate is a tert-butyl ester derivative used as a biochemical assay reagent and synthetic building block in research applications. It has CAS number 234082-00-9, molecular formula C18H34O4, and molecular weight 314.46 g/mol. Storage recommendations include room temperature storage for up to 3 years; in solvent, store at -80°C for 2 years or -20°C for 1 year.

  • CAS number: 234082-00-9
  • Molecular formula: C18H34O4
  • Molecular weight: 314.46 g/mol
  • Intended use: biochemical assay reagents
  • Storage: room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.