Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl hydrogen tetradecanedioate | 234082-00-9 | MFCD23136040 | 99.9% | 314.46 g/mol | C18H34O4 | 10 G
SDP

Catalog No. 5000210400
Encompass_Preferred

Tert-butyl hydrogen tetradecanedioate is a tert-butyl ester of tetradecanedioic acid used as a short linker and intermediate in organic synthesis and biochemical assays. It is supplied for laboratory research and characterized by high purity and defined molecular properties.

  • High purity: 99.9% (HPLC)
  • Molecular weight: 314.46 g/mol
  • Chemical formula: C18H34O4
  • Typical applications: linker and intermediate in organic synthesis and biochemical assays
  • Common lab quantity: 10 G

Catalog No. 50-002-10400 Supplier Medchemexpress LLC Supplier No. HYW07495310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tert-butyl hydrogen tetradecanedioate is a tert-butyl ester of tetradecanedioic acid used as a short linker and intermediate in organic synthesis and biochemical assays. It is supplied for laboratory research and characterized by high purity and defined molecular properties.

  • High purity: 99.9% (HPLC)
  • Molecular weight: 314.46 g/mol
  • Chemical formula: C18H34O4
  • Typical applications: linker and intermediate in organic synthesis and biochemical assays
  • Common lab quantity: 10 G

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.