Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butyl hydroperoxide solution

Catalog No. 41666525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-666-525ML 25 mL
41-666-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-666-525ML Supplier Sigma Aldrich Supplier No. 41666525ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butyl hydroperoxide is an organic peroxide, used in oxidation processes.tert-Butyl hydroperoxide may be used in:osmium catalyzed vicinal hydroxylation of olefins under alkaline conditions catalytic asymmetric oxidation of sulfides to sulfoxides using binaphthol as a chiral auxiliaryoxidation of dibenzothiophenes

Specifications

CAS 75-91-2
Density 0.808 g/mL (at 25°C)
Molecular Formula C4H10O2
Linear Formula (CH3)3 COOH
Refractive Index n20/D 1.402
Quantity 25 mL
MDL Number MFCD00002130
Synonym 1,1-Dimethylethyl hydroperoxide; 2-Hydroperoxy-2-methylpropane; TBHP
Molecular Weight (g/mol) 90.12
Percent Purity 55.4-65.1% (Thiosulphate, titration)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.