Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butyl isocyanate

Catalog No. 1444525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-445-25G 5 g
14-445-2100G 100 g
14-445-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-445-25G Supplier Sigma Aldrich Supplier No. 1444525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tert-butyl isocyanate undergoes [2+2] cycloaddition reaction with phosphagermaallene to yield 1-oxa-2-germacyclobutane. it also undergoes cycladdition reaction with molybdenum dioxo bis(aryloxide) complex to form novel 16-electron molybdenum oxo-imido bis(aryloxide) complex. it reacts with aluminum dihydride in the presence of trace amounts of water to yield terminal hydroxide containing dinuclear alumoxane.tert-butyl isocyanate can be used:
  • In the preparation of isocyanate-treated graphite oxides (igos) which can be exfoliated in polar aprotic solvents to obtain graphene oxide nanoplatelets
  • To modify metal-organic frameworks, postsynthetically, to obtain microporous urea-functionalized frameworks
  • To the synthesis of terminal hydroxide containing dinuclear alumoxane from aluminum dihydride

Specifications

CAS 1609-86-5
Density 0.868 g/mL (at 25°C (literature))
Boiling Point 85°C to 86°C (lit.)
Molecular Formula C5H9NO
Linear Formula (CH3)3 CN=C=O
Refractive Index n20/D 1.386 (literature)
MDL Number MFCD00002035
Quantity 5 g
Synonym 2-isocyanato-2-methylpropane
Molecular Weight (g/mol) 99.13
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.