Learn More
tert-Butyl Methyl Ether
Specifications
Specifications
| CAS | 1634-04-4 |
| Quantity | 1 x 1 mL |
| Packaging | Ampule |
| Molecular Formula | C5H12O |
| Synonym | tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane |
| InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
| SMILES | CC(C)(C)OC |
| IUPAC Name | 2-methoxy-2-methylpropane |
| Molecular Weight (g/mol) | 88.15 |
| PubChem CID | 15413 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.