Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl methyl ether, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019145 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9145 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9145 Supplier MilliporeSigma Supelco Supplier No. 086035MLF
Add to Cart
Add to Cart

tert-Butyl methyl ether or methyl tert-butyl ether (MTBE) is an oxygenated compound.. It is commonly used as a gasoline additive in fuel. Though it is known to control air pollution, it is a potential groundwater pollutant.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 705°F
CAS 1634-04-4
Vapor Density 3.1 (vs. air)
Density 0.74 g/mL (at 25°C)
Flash Point -28°C - Closed Cup
Linear Formula (CH3)3 COCH3
MDL Number MFCD00008812
Boiling Point 55°C to 56°C
Synonym MTBE; Methyl tert-butyl ether
Type Analytical Standard
Quantity 5 mL
Formula Weight 88.15
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.