Learn More
Sigma Aldrich Fine Chemicals Biosciences tert Butyl methyl ether AC
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 44380818LCS
Encompass
tert-Butyl methyl ether (methyl tert-butyl ether MTBE) is a gasoline additive. Its oxidative degradation by propane-oxidizing bacterial strains has been tested. Kinetics of its heat-mediated biodegradation has been investigated under various reaction conditions. Its biodegradation followed a pseudo-first-order decay model and its pseudo-first-order rate constants were evaluated. Its extraction from water has been proposed by employing an organic polymeric compound Nafion. MTBE can be synthesized by acid-catalyzed reaction between methanol and isobutene. A study suggests that the addition of MTBE increases the number of active sites during polymerization of propene by stopped-flow method. This solvent meets ACS (American Chemical Society) specifications and can be used for processes requiring strict quality conditions such as analytical testing.
Catalog No. 50-290-3738