Learn More
Sigma Aldrich Fine Chemicals Biosciences tert Butyl methyl ether an1L
Catalog No. 502897395
Encompass
tert-Butyl methyl ether is a commonly used organic solvent that can be synthesized by acid catalyzed reaction between methanol and isobutene. It is an effective alternative to lead containing additives for enhancing the octane rating of gasoline.
Catalog No. 50-289-7395 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3069756X1L
tert-Butyl methyl ether is a commonly used organic solvent that can be synthesized by acid catalyzed reaction between methanol and isobutene. It is an effective alternative to lead containing additives for enhancing the octane rating of gasoline.