Learn More
tert-Butyl methyl ether CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™
Shop All Honeywell Research Chemicals ProductsDescription
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Density | 0.74g/mL at 25°C |
| Quantity | 2.5 L |
| Refractive Index | n20/D 1.369 |
| Linear Formula | (CH3)3COCH3 |
| Beilstein | 1730942 |
| Synonym | tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane |
| Boiling Point | 55°C to 56°C |
| SMILES | CC(C)(C)OC |
| Formula Weight | 88.15g/mol |
| Percent Purity | ≥99.8% |
| Show More |
Safety and Handling
| Hazard Statement | P210 |
| Precautionary Statement | H225-H315 |
| EINECSNumber | 216-653-1 |
| RTECSNumber | KN5250000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.