Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl methyl ether CHROMASOLV™, for HPLC, ≥99.8%, Solstice

Catalog No. 6001647 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-016-47 4 x 2.5 L
60-016-44 1 L
60-016-45 2.5 L
60-048-236 4 x 4 L
60-016-43 100 mL
60-016-48 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-016-47 Supplier Solstice Supplier No. 348754X2.5L
Only null left
Add to Cart
Add to Cart

CHROMASOLV™, for HPLC,≥99.8%

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane

Specifications

Density 0.74g/mL at 25°C
Quantity 4 x 2.5 L
Refractive Index n20/D 1.369
Linear Formula (CH3)3COCH3
Beilstein 1730942
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
Boiling Point 55°C to 56°C
SMILES CC(C)(C)OC
Formula Weight 88.15g/mol
Percent Purity ≥99.8%
Grade CHROMASOLV
Chemical Name or Material tert-Butyl methyl ether, for HPLC
Show More Show Less
Hazard Statement P210
Precautionary Statement H225-H315
EINECSNumber 216-653-1
RTECSNumber KN5250000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.