Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl methyl ether, For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™
GSA_VA

Catalog No. M1019951000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1019951000 1 L Glass Bottle
M1019952500 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1019951000 Supplier MilliporeSigma Supplier No. 1.01995.1000
Add to Cart
Add to Cart

For gas chromatography ECD and FID SupraSolv™

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -108.6°C
Color Colorless
Density 0.74g/cm3 at 20°C
Quantity 1 L
Flash Point −28°C (−18.4°F)
Ignition Point 460°C (860°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 268 hPa (20°C)
UN Number 2398
Boiling Point 55.3°C
Solubility Information Soluble in water: 42g/L at 20°C
Formula Weight 88.15
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H315 Causes skin irritation.
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. Protected from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.