Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl methyl ether, Solstice

Catalog No. 60048590 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
202.7 L
Packaging:
Glass Bottle
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-590 202.7 L Metal Drum
60-048-589 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-590 Supplier Solstice Supplier No. 20256202.7L
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Puriss. p.a., ≥99.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
IUPAC Name 2-methoxy-2-methylpropane
SMILES COC(C)(C)C

Specifications

Quantity 202.7 L
Packaging Metal Drum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.