Learn More
Sigma Aldrich Fine Chemicals Biosciences tert-Butyl methyl ether HP1L
Catalog No. 502897884
Encompass
tert-Butyl methyl ether is a gasoline additive. Its oxidative degradation by propane-oxidizing bacterial strains has been tested. Kinetics of its heat-assisted persulfate oxidation has been investigated at various pH temperature oxidant concentration and ionic strength levels. Its biodegradation followed a pseudo-first-order decay model and its pseudo-first-order rate constants were evaluated. MTBE can be synthesized by acid catalyzed reaction between methanol and isobutene. A study suggests that the addition of MTBE increases the number of active sites during polymerization of propene by stopped-flow method.
Catalog No. 50-289-7884 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6505601L
tert-Butyl methyl ether is a gasoline additive. Its oxidative degradation by propane-oxidizing bacterial strains has been tested. Kinetics of its heat-assisted persulfate oxidation has been investigated at various pH temperature oxidant concentration and ionic strength levels. Its biodegradation followed a pseudo-first-order decay model and its pseudo-first-order rate constants were evaluated. MTBE can be synthesized by acid catalyzed reaction between methanol and isobutene. A study suggests that the addition of MTBE increases the number of active sites during polymerization of propene by stopped-flow method.