Learn More
Sigma Aldrich Fine Chemicals Biosciences tert-Butyl methyl ether su1L
Catalog No. 502899372
Encompass
tert-Butyl methyl ether (methyl tert-butyl ether MTBE) is a gasoline additive. Its oxidative degradation by propane-oxidizing bacterial strains has been tested. Kinetic studies of its heat-assisted persulfate oxidation at various pH temperature oxidant concentration and ionic strength levels suggests that reaction exhibits a pseudo-first-order decay model.
Catalog No. 50-289-9372 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348751L
tert-Butyl methyl ether (methyl tert-butyl ether MTBE) is a gasoline additive. Its oxidative degradation by propane-oxidizing bacterial strains has been tested. Kinetic studies of its heat-assisted persulfate oxidation at various pH temperature oxidant concentration and ionic strength levels suggests that reaction exhibits a pseudo-first-order decay model.