Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Methyl Iminodicarboxylate 98.0+%, TCI America™
SDP

Catalog No. I05105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05105G Supplier TCI America Supplier No. I05105G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Precursor of Modified Gabriel reagent for the introduction of Boc-amino groups in chemical synthesis.

Chemical Identifiers

CAS 66389-76-2
Molecular Formula C7H13NO4
Molecular Weight (g/mol) 175.18
MDL Number MFCD01321176
InChI Key BPECCYFUJCNQOR-UHFFFAOYSA-N
Synonym Iminodicarboxylic Acid tert-Butyl Methyl Ester
PubChem CID 11480754
IUPAC Name tert-butyl N-(methoxycarbonyl)carbamate
SMILES COC(=O)NC(=O)OC(C)(C)C

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 5 g
Formula Weight 175.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl Methyl Iminodicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.