Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl MethyleEther, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog No. AC610271000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC610271000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC610271000 Supplier Thermo Scientific Chemicals Supplier No. 610271000
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Quantity 100 mL
Identification Pass Test
Assay Percent Range 99.00%
Vapor Pressure Vapor Pressure: 268mbar at 20°C
Linear Formula (CH3)3COCH3
Water 50ppm max.
Beilstein 01, 381
Fieser 04,333
Merck Index 15, 6106
Formula Weight 88.15
Percent Purity ≥99%
Grade Extra Dry
Chemical Name or Material tert-Butyl methyleether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS P Statement
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.