Learn More
Medchemexpress LLC Tert-butyl N-(1H-indol-4-yl)carbamate | 819850-13-0 | MFCD04973303 | 95.0-98.0% | 232.28 g/mol | C13H16N2O2 | 100 MG
SDP
Catalog No. 5000282958
Encompass_Preferred
tert-Butyl 1H-indol-4-ylcarbamate is a synthetic intermediate used in pharmaceutical and medicinal chemistry research. Supplied as a powder for laboratory use and intended for research use only, it serves as a building block in small-molecule synthesis and related workflows.
  • Used as a synthetic intermediate in pharmaceutical synthesis.
  • Supplied as a powder suitable for laboratory handling.
  • Available in multiple pack sizes, including 100 mg, 250 mg, 500 mg, and 1 g.
  • Identified by CAS 819850-13-0 and molecular formula C13H16N2O2.
  • Storage: powder stable at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Product datasheet available; purity and batch details provided by suppliers.
Catalog No. 50-002-82958 Supplier Medchemexpress LLC Supplier No. HYW129533100MG
tert-Butyl 1H-indol-4-ylcarbamate is a synthetic intermediate used in pharmaceutical and medicinal chemistry research. Supplied as a powder for laboratory use and intended for research use only, it serves as a building block in small-molecule synthesis and related workflows.
  • Used as a synthetic intermediate in pharmaceutical synthesis.
  • Supplied as a powder suitable for laboratory handling.
  • Available in multiple pack sizes, including 100 mg, 250 mg, 500 mg, and 1 g.
  • Identified by CAS 819850-13-0 and molecular formula C13H16N2O2.
  • Storage: powder stable at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Product datasheet available; purity and batch details provided by suppliers.