Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl N-[2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]ethyl]carbamate | 811442-84-9 | MFCD16619339 | >=98.0% | 336.42 g/mol | C15H32N2O6 | 10 G
SDP

Catalog No. 5000204273
Encompass_Preferred

Boc-NH-PEG4-CH2CH2NH2 is a Boc-protected PEG4 diamine linker used as a cleavable spacer in antibody-drug conjugate (ADC) synthesis and as a linker in PROTAC and other bioconjugation chemistries.

  • Boc-protected terminal amine for orthogonal deprotection.
  • Peg4 spacer provides a short, flexible linkage for conjugation.
  • Tert-butyl N-[2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]ethyl]carbamate is the preferred IUPAC name.
  • High purity (≥98.0%).
  • Molecular weight 336.42 g/mol and molecular formula C15H32N2O6.
  • Storage at 4°C, protected from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in multiple pack sizes, including 10 G.

Catalog No. 50-002-04273 Supplier Medchemexpress LLC Supplier No. HYW00835210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-NH-PEG4-CH2CH2NH2 is a Boc-protected PEG4 diamine linker used as a cleavable spacer in antibody-drug conjugate (ADC) synthesis and as a linker in PROTAC and other bioconjugation chemistries.

  • Boc-protected terminal amine for orthogonal deprotection.
  • Peg4 spacer provides a short, flexible linkage for conjugation.
  • Tert-butyl N-[2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]ethyl]carbamate is the preferred IUPAC name.
  • High purity (≥98.0%).
  • Molecular weight 336.42 g/mol and molecular formula C15H32N2O6.
  • Storage at 4°C, protected from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in multiple pack sizes, including 10 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.