Learn More
Medchemexpress LLC Tert-butyl N-[2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]ethyl]carbamate | 811442-84-9 | MFCD16619339 | ≥98.0% | 336.42 g/mol | C15H32N2O6 | 5 G
SDP
Catalog No. 5000203070
Encompass_Preferred
Boc-NH-PEG4-CH2CH2NH2 is a Boc-protected, 5-unit polyethylene glycol diamine linker used as a cleavable linker in the synthesis of antibody-drug conjugates (ADCs) and as a PEG-based linker for PROTAC and other chemical biology applications.
  • Boc-protected PEG4 diamine structure.
  • Cleavable 5-unit PEG linker suitable for ADC synthesis.
  • High purity supported by COA (NMR ≥98.0%).
  • Available in multiple package sizes, including 5 G.
  • Storage: protect from light; storage stability varies with solvent.
Catalog No. 50-002-03070 Supplier Medchemexpress LLC Supplier No. HYW0083525G
Boc-NH-PEG4-CH2CH2NH2 is a Boc-protected, 5-unit polyethylene glycol diamine linker used as a cleavable linker in the synthesis of antibody-drug conjugates (ADCs) and as a PEG-based linker for PROTAC and other chemical biology applications.
  • Boc-protected PEG4 diamine structure.
  • Cleavable 5-unit PEG linker suitable for ADC synthesis.
  • High purity supported by COA (NMR ≥98.0%).
  • Available in multiple package sizes, including 5 G.
  • Storage: protect from light; storage stability varies with solvent.