Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich tert-Butyl N-(2,3-dihydroxypropyl)carbamate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Please note that sigma-aldrich provides this product to early discovery researchers as part of a collection of unique chemicals
- Sigma-aldrich does not collect analytical data for this product
- Buyer assumes responsibility to confirm product identity and/or purity
- All sales are final.notwithstanding any contrary provision contained in sigma-aldrich's standard terms and conditions of sale or an agreement between sigma-aldrich and buyer, sigma-aldrich sells this product as-is and makes no representation or warranty whatsoever with respect to this product, including any (a) warranty of merchantability; (b) warranty of fitness for a particular purpose; or (c) warranty against infringement of intellectual property rights of a third party; whether arising by law, course of dealing, course of performance, usage of trade or otherwise.product of chembridge corp
Spécifications
Spécifications
| CAS | 137618-48-5 |
| Melting Point | 60°C to 63°C (lit.) |
| Molecular Formula | C8H17NO4 |
| Linear Formula | HOCH2CH(OH)CH2NHCO2C(CH3)3 |
| MDL Number | MFCD01321265 |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 191.22 |
| Percent Purity | 97% |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'