Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl N-(2-cyanoethyl)carbamate, 97%, Thermo Scientific™

Catalog No. SPB08399EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SPB08399EA 10 g Amber Glass Bottle
SPB08399DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SPB08399EA Supplier Thermo Scientific Chemicals Supplier No. SPB08399EA
Add to Cart

Chemical Identifiers

CAS 53588-95-7
Molecular Formula C8H14N2O2
Molecular Weight (g/mol) 170.21
MDL Number MFCD02180880
InChI Key NORLFIHQJFOIGS-UHFFFAOYSA-N
Synonym tert-butyl n-2-cyanoethyl carbamate, tert-butyl 2-cyanoethyl carbamate, n-boc-2-cyano-ethylamine, 2-cyano-ethyl-carbamic acid tert-butyl ester, t-butyl n-2-cyanoethyl carbamate, tert-butoxy-n-2-cyanoethyl carboxamide, 1,1-dimethylethyl 2-cyanoethyl carbamate, 3-tert-butoxycarbonyl amino propanenitrile, carbamicacid,n-2-cyanoethyl-,1,1-dimethylethylester
PubChem CID 2735664
IUPAC Name tert-butyl N-(2-cyanoethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCC#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)3COC(O)NH(CH2)2CN
Quantity 10 g
Formula Weight 170.21
Percent Purity 97%
Chemical Name or Material Tert-butyl N-(2-cyanoethyl)carbamate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.