Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl N-(2-Hydroxypropyl)carbamate 98.0+%, TCI America™
SDP

Catalog No. B24185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24185G 5 g
B241825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24185G Supplier TCI America Supplier No. B24185G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 95656-86-3
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD02093569
InChI Key YNJCFDAODGKHAV-UHFFFAOYNA-N
Synonym 1-(Boc-amino)-2-propanol, 1-(tert-Butoxycarbonylamino)-2-propanol, N-(2-Hydroxypropyl)carbamic Acid tert-Butyl Ester
PubChem CID 9920508
IUPAC Name tert-butyl N-(2-hydroxypropyl)carbamate
SMILES CC(O)CNC(=O)OC(C)(C)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 175.23
Percent Purity ≥98.0% (N)
Grade Research
Physical Form Liquid
Chemical Name or Material tert-Butyl N-(2-Hydroxypropyl)carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.