Learn More
Medchemexpress LLC Tert-butyl N-[3-(piperazin-1-yl)propyl]carbamate | 874831-60-4 | MFCD06808577 | >97.0% | 243.35 | C12H25N3O2 | 10 G

Supplier: Medchemexpress LLC HYW08845610G
tert-Butyl N-[3-(piperazin-1-yl)propyl]carbamate is a chemical linker and synthetic intermediate used in PROTAC development; it is documented as a building block for synthesis of the degrader TrimTAC1 and is supplied as a liquid reagent for research use.
- Used as a PROTAC linker and synthetic intermediate.
- CAS number: 874831-60-4.
- Molecular formula: C12H25N3O2.
- Molecular weight: 243.35 g/mol.
- Purity: >97.0%.
- Physical form: liquid; colorless to light yellow.
- Storage: store at 4°C and protect from light; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
- Available in multiple package sizes, including 10 G.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.