Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl N-[3-(piperazin-1-yl)propyl]carbamate | 874831-60-4 | MFCD06808577 | >=97.0% | 243.35 | C12H25N3O2 | 250 MG
SDP

Catalog No. 5000209305
Encompass_Preferred

tert-Butyl N-[3-(piperazin-1-yl)propyl]carbamate is a tert-butyl carbamate protected piperazine-containing linker used as a building block in medicinal chemistry and targeted protein degradation research. It is provided as a high-purity reagent suitable for synthesis of bifunctional molecules and linkers.

  • high purity suitable for research use
  • liquid appearance, colorless to light yellow
  • molecular weight 243.35
  • chemical formula C12H25N3O2
  • available in multiple pack sizes, including 250 mg
  • storage: protect from light and store refrigerated; in solvent, store at -80°C for long-term stability

Catalog No. 50-002-09305 Supplier Medchemexpress LLC Supplier No. 250MGHYW088456
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl N-[3-(piperazin-1-yl)propyl]carbamate is a tert-butyl carbamate protected piperazine-containing linker used as a building block in medicinal chemistry and targeted protein degradation research. It is provided as a high-purity reagent suitable for synthesis of bifunctional molecules and linkers.

  • high purity suitable for research use
  • liquid appearance, colorless to light yellow
  • molecular weight 243.35
  • chemical formula C12H25N3O2
  • available in multiple pack sizes, including 250 mg
  • storage: protect from light and store refrigerated; in solvent, store at -80°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.