Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl N-(3-thienyl)carbamate, 97%, Thermo Scientific™

Catalog No. CC00092CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC00092CB 250 mg Amber Glass Bottle
CC00092DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC00092CB Supplier Thermo Scientific Chemicals Supplier No. CC00092CB
Add to Cart

Chemical Identifiers

CAS 19228-91-2
Molecular Formula C9H13NO2S
Molecular Weight (g/mol) 199.268
MDL Number MFCD01928808
InChI Key PRWYQCYSADTIBZ-UHFFFAOYSA-N
Synonym tert-butyl thiophen-3-ylcarbamate, tert-butyl n-3-thienyl carbamate, tert-butyl 3-thienylcarbamate, tert-butyl n-thiophen-3-yl carbamate, tert-butoxy-n-3-thienyl carboxamide, carbamic acid,n-3-thienyl-, 1,1-dimethylethyl ester, n-boc-3-aminothiophene, tert-butyl thien-3-ylcarbamate, 3-aminothiophene, n-boc protected
PubChem CID 736476
IUPAC Name tert-butyl N-thiophen-3-ylcarbamate
SMILES CC(C)(C)OC(=O)NC1=CSC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.27
Percent Purity 97%
Chemical Name or Material Tert-butyl N-(3-thienyl)carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.