Learn More
Medchemexpress LLC Tert-butyl N-[(3R)-1-[[2-[[4-(4-morpholin-4-yl-7H-pyrrolo[2,3-d]pyrimidin-6-yl)phenyl]carb | 2448173-47-3 | MFCD34574791 | 98.8% | 612.72 g/mol | C33H40N8O4 | 5 MG
SDP
Catalog No. 5000263118
Encompass_Preferred
Menin-MLL inhibitor 20 is an irreversible small-molecule inhibitor of the menin-MLL protein-protein interaction with reported antitumor activity. Supplied as a quality-controlled research reagent, it is provided in solid and solution formats for in vitro and preclinical studies; documentation includes a product datasheet and a certificate of analysis.
  • Irreversible menin-MLL interaction inhibitor with reported antitumor activity.
  • High purity (98.8%) suitable for research applications.
  • Molecular formula C33H40N8O4, molecular weight 612.72 g/mol.
  • Supplied as a solid with main pack sizes from 5 mg to 100 mg and a solution format available.
  • Includes datasheet and certificate of analysis for quality verification.
  • Intended for in vitro and preclinical research use only.
This item is not returnable. View return policy
Catalog No. 50-002-63118 Supplier Medchemexpress LLC Supplier No. HY1287985MG
Menin-MLL inhibitor 20 is an irreversible small-molecule inhibitor of the menin-MLL protein-protein interaction with reported antitumor activity. Supplied as a quality-controlled research reagent, it is provided in solid and solution formats for in vitro and preclinical studies; documentation includes a product datasheet and a certificate of analysis.
  • Irreversible menin-MLL interaction inhibitor with reported antitumor activity.
  • High purity (98.8%) suitable for research applications.
  • Molecular formula C33H40N8O4, molecular weight 612.72 g/mol.
  • Supplied as a solid with main pack sizes from 5 mg to 100 mg and a solution format available.
  • Includes datasheet and certificate of analysis for quality verification.
  • Intended for in vitro and preclinical research use only.