Learn More
Medchemexpress LLC Tert-butyl N-(5-hydroxypentyl)carbamate | 75178-90-4 | MFCD01862955 | >97.0% | 203.28 g·mol⁻1 | C10H21NO3 | 1 G
SDP
Catalog No. 5000299749
Encompass_Preferred
5-(Boc-amino)-1-pentanol is a Boc-protected 5-aminopentanol used as a versatile intermediate and linker in organic and medicinal chemistry. It contains a tert-butoxycarbonyl protected amine and a primary alcohol, enabling selective deprotection and further functionalization for synthesis of peptides, small molecules, and conjugates.
  • Boc-protected amine enables selective deprotection workflows.
  • Primary alcohol allows further derivatization and coupling reactions.
  • High purity suitable for synthetic and medicinal chemistry applications.
  • Defined storage stability in solvent and bulk formats.
  • Useful as a linker for conjugation and building-block synthesis.
Catalog No. 50-002-99749 Supplier Medchemexpress LLC Supplier No. HYW0397291G
5-(Boc-amino)-1-pentanol is a Boc-protected 5-aminopentanol used as a versatile intermediate and linker in organic and medicinal chemistry. It contains a tert-butoxycarbonyl protected amine and a primary alcohol, enabling selective deprotection and further functionalization for synthesis of peptides, small molecules, and conjugates.
  • Boc-protected amine enables selective deprotection workflows.
  • Primary alcohol allows further derivatization and coupling reactions.
  • High purity suitable for synthetic and medicinal chemistry applications.
  • Defined storage stability in solvent and bulk formats.
  • Useful as a linker for conjugation and building-block synthesis.