Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl N-(6-bromohexyl)carbamate | 142356-33-0 | MFCD06201019 | 98.0% | 280.20 g/mol | C11H22BrNO2 | 5 G
SDP

Catalog No. 5000203220
Encompass_Preferred

Boc-NH-C6-Br is a non-cleavable linker used in the synthesis of antibody-drug conjugates (ADCs). It provides a stable, non-cleavable connection between an antibody and a cytotoxic payload and is supplied for research use only.

  • Non-cleavable linker for ADC synthesis.
  • Purity 98.0%.
  • Molecular formula C11H22BrNO2; molecular weight 280.20 g/mol.
  • Appearance colorless to off-white; form solid-liquid mixture.
  • Storage: pure form -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended for research use only; not for human or veterinary applications.

Catalog No. 50-002-03220 Supplier Medchemexpress LLC Supplier No. HYW0115615G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-NH-C6-Br is a non-cleavable linker used in the synthesis of antibody-drug conjugates (ADCs). It provides a stable, non-cleavable connection between an antibody and a cytotoxic payload and is supplied for research use only.

  • Non-cleavable linker for ADC synthesis.
  • Purity 98.0%.
  • Molecular formula C11H22BrNO2; molecular weight 280.20 g/mol.
  • Appearance colorless to off-white; form solid-liquid mixture.
  • Storage: pure form -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended for research use only; not for human or veterinary applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.