Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl N-(Benzyloxy)carbamate 98.0+%, TCI America™
SDP

Catalog No. B17995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17995G 5 g
B179925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17995G Supplier TCI America Supplier No. B17995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79722-21-7
Molecular Formula C12H17NO3
Molecular Weight (g/mol) 223.272
MDL Number MFCD00191873
InChI Key MZNBNPWFHGWAGH-UHFFFAOYSA-N
Synonym N-(Benzyloxy)carbamic Acid tert-Butyl Ester
PubChem CID 736159
IUPAC Name tert-butyl N-phenylmethoxycarbamate
SMILES CC(C)(C)OC(=O)NOCC1=CC=CC=C1

Specifications

Melting Point 48°C
Color White-Yellow
Quantity 5 g
Formula Weight 223.27
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl N-(Benzyloxy)carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.