Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butyl nitrite

Catalog No. 23538525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-538-525ML 25 mL
23-538-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-538-525ML Supplier Sigma Aldrich Supplier No. 23538525ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tert-butyl nitrite (tbn) is an efficient no source. tbn participates in photocatalyzed conversion of aryl- and heteroarylamines to selenides. it also participates in radical multifunctionalization reactions of aliphatic alkenes.tert-butyl nitrite has been used as reagent:
  • For diazotization and nitros-íation of alcohols, thiols, amines and cycloalkanes
  • For the preparation of aryl azides from aryl aminesremainder 2-methyl-2-propanol

Specifications

CAS 540-80-7
Density 0.867 g/mL (at 25°C (literature))
Boiling Point 61°C to 63°C (lit.)
Molecular Formula C4H9NO2
Refractive Index n20/D 1.368 (literature)
Linear Formula (CH3)3 CONO
MDL Number MFCD00002055
Quantity 25 mL
Molecular Weight (g/mol) 103.12
Percent Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.