Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butyl nitrite

Catalog No. 20370500MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-370-500MLF 500 mL
20-370-100MLF 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-370-500MLF Supplier Sigma Aldrich Supplier No. 20370500MLF
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for nitrosations, kinetics; Clean source of Me3C radicals by near-UV photolysis

Specifications

CAS 540-80-7
Density 0.864 g/mL at 20°C, 0.867 g/mL (at 25°C (literature))
Boiling Point 61°C to 63°C (lit.)
Molecular Formula C4H9NO2
Refractive Index n20/D 1.368 (literature); n20/D 1.368
Linear Formula (CH3)3 CONO
Quantity 500 mL
MDL Number MFCD00002055
Molecular Weight (g/mol) 103.12
Percent Purity ≥90% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.