Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tert-Butyl P,P-Dimethylphosphonoacetate

Catalog No. 7952225ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-522-25ML 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 79-522-25ML Supplier Sigma Aldrich Supplier No. 7952225ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Preparation of phosphonate terminated PPH dendrimers as anti-HIV-1 agentsPreparation of monodehydro diketopiperazines from ketoacyl amino acid amides via acid-catalyzed cyclizationPreparation of +a,+a-unsaturated esters for use in the guanidine-catalyzed oxa-Michael additionPreparation of myxopyronin B and desmethyl myxopyronin B analogs, with antibacterial activity and inhibitory activity against bacterial RNA polymeraseAllylation and subsequent ring-closing metathesis in presence of Grubbs'sa catalyst or intramolecular rhodium-catalyzed cyclopropanation reactionsHorner-Wittig reagent giving preferentially (E)-+a,+a-unsat. esters which are mildly saponified with acid. Synthesis of derivatives by reductive alkylation

Specifications

CAS 169243-86-1
Molecular Formula C18H20N2O2S2
Refractive Index n20/D 1.434
Linear Formula C18H20N2O2S2
MDL Number MFCD00083161
Quantity 50 mg
Synonym tert-Butyl ((oxo)dimethoxyphosphino)acetate; tert-Butyl (dimethoxyphosphinyl)acetate; Dimethyl (tert-butoxycarbonyl)methylphosphonate
Molecular Weight (g/mol) 360.49 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.