Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tert-Butyl p-Toluate | 13756-42-8 | C₁₂H₁₆O₂ | 50 G
SDP

Catalog No. 5000434646
Encompass_Preferred

tert-Butyl p-Toluate (tert-Butyl p-methylbenzoate) is a synthetic intermediate useful for pharmaceutical synthesis. This product is for research use only and has not been fully validated for medical applications.

  • A synthetic intermediate
  • Useful for pharmaceutical synthesis
  • Molecular weight: 192.25
  • Chemical formula: C₁₂H₁₆O₂
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Store at room temperature for 3 years
  • For storage in solvent: -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-34646 Supplier Medchemexpress LLC Supplier No. HY7654850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl p-Toluate (tert-Butyl p-methylbenzoate) is a synthetic intermediate useful for pharmaceutical synthesis. This product is for research use only and has not been fully validated for medical applications.

  • A synthetic intermediate
  • Useful for pharmaceutical synthesis
  • Molecular weight: 192.25
  • Chemical formula: C₁₂H₁₆O₂
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Store at room temperature for 3 years
  • For storage in solvent: -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.