Learn More
Medchemexpress LLC tert-Butyl p-Toluate | 13756-42-8 | C12H16O2 | 25 G
SDP
Catalog No. 5000359186
Encompass_Preferred
tert-Butyl p-Toluate is a synthetic intermediate useful for pharmaceutical synthesis. It is also known as tert-Butyl p-methylbenzoate.
  • Useful for pharmaceutical synthesis
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years
  • Store in solvent at -20°C for 1 year
Catalog No. 50-003-59186 Supplier Medchemexpress LLC Supplier No. 25GHY76548
tert-Butyl p-Toluate is a synthetic intermediate useful for pharmaceutical synthesis. It is also known as tert-Butyl p-methylbenzoate.
  • Useful for pharmaceutical synthesis
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years
  • Store in solvent at -20°C for 1 year