Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl sarcosinate hydrochloride | 136088-69-2 | MFCD00080936 | 98.0% | 181.66 | C7H16ClNO2 | 500 G
SDP

Catalog No. 5000300811
Encompass_Preferred

H-Sar-OtBu·HCl (tert-butyl sarcosinate hydrochloride, CAS 136088-69-2) is the hydrochloride salt of the tert-butyl ester of sarcosine. Supplied as a white to off-white solid, it serves as a protected amino-acid reagent and intermediate for peptide and small-molecule synthesis.

  • White to off-white solid
  • Chemical formula C7H16ClNO2; molecular weight 181.66
  • Purity 98.0%
  • Store sealed at 4°C and protect from moisture
  • Used as a protected sarcosine building block in peptide synthesis
  • Available in laboratory sizes suitable for synthetic workflows

Catalog No. 50-003-00811 Supplier Medchemexpress LLC Supplier No. HYW018062500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Sar-OtBu·HCl (tert-butyl sarcosinate hydrochloride, CAS 136088-69-2) is the hydrochloride salt of the tert-butyl ester of sarcosine. Supplied as a white to off-white solid, it serves as a protected amino-acid reagent and intermediate for peptide and small-molecule synthesis.

  • White to off-white solid
  • Chemical formula C7H16ClNO2; molecular weight 181.66
  • Purity 98.0%
  • Store sealed at 4°C and protect from moisture
  • Used as a protected sarcosine building block in peptide synthesis
  • Available in laboratory sizes suitable for synthetic workflows

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.