Learn More
eMolecules​ 134003-83-1 | tert-butyl(1R,4R)-5-benzyl-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate | MFCD28411213 | 1g
Supplier:  eMolecules​ 599170512
Encompass_Preferred
Medchem Express | CC-671 | 5mg | 446259389 | HY-108709 | 1618658-88-0 | 512.570 | C28H28N6O4
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool<|a>
Catalog No. 50-214-4868