Learn More
Medchemexpress LLC Tert-butylamine borane | 7337-45-3 | MFCD00075635 | 99.0% | 83.95 g/mol | C4H11BN | 5 G
SDP
Catalog No. 5000283508
Encompass_Preferred
Tert-butylamine borane is a mild amine-borane reducing agent supplied as the tert-butylamine-borane complex. It is used in organic synthesis for selective reduction of aldehydes and ketones to the corresponding alcohols, and offers a more easily handled alternative to borane-ether solutions for laboratory-scale reductions.
  • Mild, chemoselective reducing agent for aldehydes and ketones.
  • Stable amine-borane complex that is easier to handle than borane-ether solutions.
  • High purity suitable for synthetic applications (99%).
  • Available in small gram quantities for laboratory use.
  • Compatible with common organic solvents under controlled conditions.
Catalog No. 50-002-83508 Supplier Medchemexpress LLC Supplier No. HYW1338975G
Tert-butylamine borane is a mild amine-borane reducing agent supplied as the tert-butylamine-borane complex. It is used in organic synthesis for selective reduction of aldehydes and ketones to the corresponding alcohols, and offers a more easily handled alternative to borane-ether solutions for laboratory-scale reductions.
  • Mild, chemoselective reducing agent for aldehydes and ketones.
  • Stable amine-borane complex that is easier to handle than borane-ether solutions.
  • High purity suitable for synthetic applications (99%).
  • Available in small gram quantities for laboratory use.
  • Compatible with common organic solvents under controlled conditions.