Learn More
Medchemexpress LLC Tert-butylamine borane | 7337-45-3 | MFCD00075635 | ≥99.0% | 86.97 g/mol | C4H14BN | 10 G
SDP
Catalog No. 5000283419
Encompass_Preferred
tert-Butylamine borane is a stable borane-amine complex used as a mild, selective reducing agent in organic synthesis. It provides controlled hydride transfer for the reduction of aldehydes and ketones to their corresponding alcohols, suitable for laboratory-scale reactions where selectivity and ease of handling are required.
  • Mild, selective hydride source for aldehyde and ketone reductions.
  • High purity (≥99.0%) for consistent reaction performance.
  • Solid, easy-to-handle complex offering improved stability over gaseous borane reagents.
  • Available in small pack sizes appropriate for bench-scale work.
  • Compatible with common organic solvents under standard reaction conditions.
Catalog No. 50-002-83419 Supplier Medchemexpress LLC Supplier No. HYW13389710G
tert-Butylamine borane is a stable borane-amine complex used as a mild, selective reducing agent in organic synthesis. It provides controlled hydride transfer for the reduction of aldehydes and ketones to their corresponding alcohols, suitable for laboratory-scale reactions where selectivity and ease of handling are required.
  • Mild, selective hydride source for aldehyde and ketone reductions.
  • High purity (≥99.0%) for consistent reaction performance.
  • Solid, easy-to-handle complex offering improved stability over gaseous borane reagents.
  • Available in small pack sizes appropriate for bench-scale work.
  • Compatible with common organic solvents under standard reaction conditions.