Learn More
Medchemexpress LLC Tert-butylamine borane | 7337-45-3 | MFCD00075635 | ≥99.0% | 86.97 g/mol | C4H14BN | 25 G
SDP
Catalog No. 5000283507
Encompass_Preferred
Tert-butylamine borane is a borane-amine complex used as a mild, chemoselective reducing agent in organic synthesis. It selectively reduces aldehydes and ketones to their corresponding alcohols and offers improved stability and handling compared with gaseous borane, making it suitable for routine laboratory reductions and small-scale synthetic applications.
  • Mild, chemoselective reducing agent for aldehydes and ketones.
  • Solid, white to off-white appearance for easy handling.
  • High purity (≥99.0%) for consistent reaction performance.
  • Soluble in common organic solvents for convenient application.
  • Stable when stored under recommended conditions to minimize degradation.
  • Available in laboratory-sized pack sizes for small-scale work.
Catalog No. 50-002-83507 Supplier Medchemexpress LLC Supplier No. HYW13389725G
Tert-butylamine borane is a borane-amine complex used as a mild, chemoselective reducing agent in organic synthesis. It selectively reduces aldehydes and ketones to their corresponding alcohols and offers improved stability and handling compared with gaseous borane, making it suitable for routine laboratory reductions and small-scale synthetic applications.
  • Mild, chemoselective reducing agent for aldehydes and ketones.
  • Solid, white to off-white appearance for easy handling.
  • High purity (≥99.0%) for consistent reaction performance.
  • Soluble in common organic solvents for convenient application.
  • Stable when stored under recommended conditions to minimize degradation.
  • Available in laboratory-sized pack sizes for small-scale work.