Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butylbenzene

Catalog No. B90602250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B90602250ML 250 mL
B906021L 1 L
B906025ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B90602250ML Supplier Sigma Aldrich Supplier No. B90602250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butylbenzene

Specifications

CAS 98-06-6
Melting Point -58°C (lit.)
Density 0.867 g/mL (at 25°C (literature))
Boiling Point 169°C (lit.)
Molecular Formula C10H14
Refractive Index n20/D 1.492 (literature)
Linear Formula C6H5C(CH3)3
MDL Number MFCD00008816
Quantity 250 mL
Synonym 2-Methyl-2-phenylpropane
Molecular Weight (g/mol) 134.22
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.