Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tert-Butylchlorodimethylsilane Solution

Catalog No. 47346450G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-346-450G 50 g
47-346-4100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 47-346-450G Supplier Sigma Aldrich Supplier No. 47346450G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tert-Butylchlorodimethylsilane Solution

Specifications

CAS 18162-48-6
Density 0.87 g/mL at 20°C, 0.866 g/mL at 25°C
Boiling Point 113°C
Molecular Formula C6H15ClSi
Linear Formula (CH3)3CSi(CH3)2Cl
Refractive Index n20/D 1.46
MDL Number MFCD00000501
Quantity 50 g
Synonym tert-Butyldimethylchlorosilane; tert-Butyldimethylsilyl chloride; TBDMSCl solution
Molecular Weight (g/mol) 150.72 g/mol
Biological Activity Central Nervous System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.