Learn More
tert-Butyldimethylchlorosilane, 97%
Description
tert-Butyldimethylchlorosilane is used to protect hydroxyl group in organic synthesis. It finds application in the synthesis of prostaglandin. It is also used as an auxiliary material for hypolipaemics such as lovastatin and simvastatin. It plays an important role in the preparation of isoxazolines N-oxides from alpha-bromonitroalkanes. It acts as a versatile protecting reagent for amines, amides and alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
tert-Butyldimethylchlorosilane is used to protect hydroxyl group in organic synthesis. It finds application in the synthesis of prostaglandin. It is also used as an auxiliary material for hypolipaemics such as lovastatin and simvastatin. It plays an important role in the preparation of isoxazolines N-oxides from alpha-bromonitroalkanes. It acts as a versatile protecting reagent for amines, amides and alcohols.
Solubility
Soluble in chloroform and ethyl acetate. Insoluble in water.
Notes
Moisture sensitive. Store in cool place. Incompatible with bases, metals, oxidizing agents and alcohols.
Specifications
Specifications
| Boiling Point | 124°C to 126°C |
| Melting Point | 86°C to 89°C |
| Quantity | 25 g |
| Linear Formula | (CH3)3CSi(CH3)2Cl |
| UN Number | UN2925 |
| Beilstein | 505999 |
| Flash Point | 22°C (72°F) |
| Solubility Information | Soluble in chloroform and ethyl acetate. Insoluble in water. |
| Formula Weight | 150.73 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H228-H314-H335 |
| Hazard Statement | GHS H Statement H228-H314-H318 Flammable solid. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: I |
| EINECSNumber | 242-042-4 |
| RTECSNumber | VV2000000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.