Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyldimethylsilyl (S)-Glycidyl Ether 95.0+%, TCI America™
SDP

Catalog No. B41115ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41115ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B41115ML Supplier TCI America Supplier No. B41115ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 123237-62-7
Molecular Formula C9H20O2Si
Molecular Weight (g/mol) 188.342
MDL Number MFCD01862148
InChI Key YANSSVVGZPNSKD-QMMMGPOBSA-N
Synonym (S)-2-[(tert-Butyldimethylsilyloxy)methyl]oxirane, (S)-1-[(tert-Butyldimethylsilyl)oxy]-2,3-epoxypropane
PubChem CID 10954307
IUPAC Name tert-butyl-dimethyl-[[(2S)-oxiran-2-yl]methoxy]silane
SMILES CC(C)(C)[Si](C)(C)OCC1CO1

Specifications

Color Yellow
Boiling Point 30°C
Quantity 5 mL
Formula Weight 188.34
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyldimethylsilyl (S)-Glycidyl Ether
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.