Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tert-Butyldimethylsilyl Trifluoromethanesulfonate

Catalog No. 2261495G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-614-95G 5 g
22-614-91G 1 g
22-614-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 22-614-95G Supplier Sigma Aldrich Supplier No. 2261495G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Together with a thiolane promotes a rapid and efficient chalcogenide-Morita-Baylis-Hillman reaction.

Specifications

CAS 69739-34-0
Density 1.151 g/mL at 25°C
Boiling Point 65°C to 67°C (12 mmHg, lit.)
Molecular Formula C7H15F3O3SSi
Refractive Index n20/D 1.385
Linear Formula CF3SO3Si(CH3)2C(CH3)3
MDL Number MFCD00000405
Quantity 5 g
Synonym TBDMS triflate; Trifluoromethanesulfonic acid tert-butyldimethylsilylester
Molecular Weight (g/mol) 264.34 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.