Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (tert-Butyldimethylsilyloxy)acetaldehyde

Catalog No. 4494581G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-945-81G 1 g
44-945-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-945-81G Supplier Sigma Aldrich Supplier No. 4494581G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • (tert-butyldimethylsilyloxy)acetaldehyde is a versatile reagent commonly used in synthetic glycobiology
  • It can act both as an aldol donor and an aldol acceptor in the stereocontrolled production of erythrose
  • It is used as an important reagent in the total synthesis of (+)-ambruticin, ()-laulimalide, ()-salinosporamide a, and (+)-leucascandrolide a

Specifications

CAS 102191-92-4
Density 0.915 g/mL (at 25°C (literature))
Boiling Point 165°C to 167°C (lit.)
Molecular Formula C8H18O2Si
Refractive Index n20/D 1.432 (literature)
Linear Formula (CH3)3 CSi(CH3)2 OCH2CHO
MDL Number MFCD01321229
Quantity 1 g
Molecular Weight (g/mol) 174.31
Percent Purity 90%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.