Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylhydroquinone Diacetate, Thermo Scientific™

Catalog No. AC403560250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC403560250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC403560250 Supplier Thermo Scientific Chemicals Supplier No. 403560250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7507-48-4
Molecular Formula C14H18O4
Quantity 25g
MDL Number MFCD00078251
Synonym tert-butylhydroquinone diacetate, 2-tert-butyl-1,4-phenylene diacetate, tert-butylhydroquinonediacetate, tert-butyl hydroquinone diacetate, tert-butylhydroquinone, diacetate, 1,4-diacetoxy-2-tert-butylbenzene, 4-acetyloxy-3-tert-butylphenyl acetate, 4-acetyloxy-2-tert-butylphenyl acetate
InChI Key SJRALGDWZXRRKL-UHFFFAOYSA-N
SMILES CC(=O)OC1=CC=C(OC(C)=O)C(=C1)C(C)(C)C
IUPAC Name 4-(acetyloxy)-2-tert-butylphenyl acetate
Molecular Weight (g/mol) 250.29
PubChem CID 82020
Formula Weight 250.29
Chemical Name or Material tert-Butylhydroquinone diacetate
Show More Show Less
EINECSNumber 231-364-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.