Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tert-Butyllithium Solution

Catalog No. 1861988L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
4 x 100 mL
8 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-619-88L 8 L
18-619-8800ML 800 mL
18-619-8100ML 4 x 100 mL
50-308-1443 4 x 25 mL
50-308-1441 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Danger
Catalog No. 18-619-88L Supplier Sigma Aldrich Supplier No. 1861988L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butyllithium solution (tert-BuLi) is an organolithium compound, used as a strong base in organic chemistry. It facilitates lithium-halogen exchange reaction and also can be employed in the deprotonation of C-H compounds and amines.tert-Butyllithium solution can be used as a strong base in the synthesis of: Alkyllithium derivatives form alkyl iodides and diiodoalkanes by halogen-lithium exchange reaction. Inorganic polymer [(LiMo3Se3)n] from [(InMo3Se3)n] by reductive intercalation method. Adducts of cinnamic acid (1,3- and 1,4-adducts) by reacting with C-C double of cinnamic acid.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 594-19-4
Density 0.652 g/mL at 25°C
Molecular Formula C4H9Li
Linear Formula (CH3)3CLi
Quantity 8 L
MDL Number MFCD00008795
Synonym Lithium-2-methyl-2-propanide; t-BuLi
Molecular Weight (g/mol) 64.06 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.