Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tert-Butylmagnesium Chloride Solution

Catalog No. 364649800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-464-9800ML 800 mL
36-464-9100ML 100 mL
36-464-925ML 4 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 36-464-9800ML Supplier Sigma Aldrich Supplier No. 364649800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butylmagnesium chloride, an organomagnesium compound, is commonly known as Grignard reagent. It is a powerful carbon nucleophile mainly used for C-C bond formation.tert-Butylmagnesium chloride (1.0 M in THF) has been used in the preparation of tert-butylsulfinyl chloride. It can be used in: Grignard metathesis reaction of 2,5-dibromo-3-hexylthiophene to form 2-bromo-5-magnesiobromo-3-hexylthiophene.A key step for the asymmetric synthesis of sulfinyl transfer agents using cinchona alkaloids quinine and quinidine as chiral auxiliaries.Storage below 25°C may cause formation of crystalline magnesium salts. Moving container to a war mLocation and ocassional gentle swirling should redissolve the solid.

Specifications

CAS 677-22-5
Density 0.931 g/mL at 25°C
Molecular Formula C4H9ClMg
Linear Formula (CH3)3CMgCl
MDL Number MFCD00000465
Quantity 800 mL
Molecular Weight (g/mol) 116.87 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.