Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylsulfinamide 97.0+%, TCI America™
SDP

Catalog No. B29265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29265G 5 g
B29261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29265G Supplier TCI America Supplier No. B29265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 146374-27-8
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.20
MDL Number MFCD01863616,MFCD05861480
InChI Key CESUXLKAADQNTB-UHFFFAOYNA-N
Synonym 2-methyl-2-propanesulfinamide, tert-butanesulfinamide, t-butylsulfinamide, tert-butylsulfinamide, 1,1-dimethylethylsulfinamide, 2-propanesulfinamide, 2-methyl, 2-methyl-propane-2-sulfinic acid amide, tert-butylsulphinamide, unii-i85yc2za8o, i85yc2za8o
PubChem CID 3382465
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(N)=O

Specifications

Melting Point 107°C
Color White
Quantity 5 g
Formula Weight 121.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylsulfinamide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.